Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPC 15199, Tocris Bioscience™

Catalog Number 0675100
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
35661-60-0
C21H23NO4
353.42
MFCD00037133
CBPJQFCAFFNICX-TWYLJJHKNA-N
fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
1549133
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid
CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
35661-60-0
C21H23NO4
353.42
MFCD00037133
CBPJQFCAFFNICX-TWYLJJHKNA-N
fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
1549133
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid
CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Increases intracellular Ca2+ levels

Anti-inflammatory agent; increases intracellular Ca2+ levels.
Formula Weight 353.42
Percent Purity >99%
Quantity 100 mg
Chemical Name or Material NPC 15199
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit