Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPDC-1 Mouse anti-Human, Clone: 160000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266632
Description
NPDC-1 Monoclonal antibody specifically detects NPDC-1 in Human samples. It is validated for Western Blot.Specifications
NPDC-1 | |
160000 | |
Western Blot | |
20MM sodium phosphate, 150MM sodium chloride, 50% glycerol, pH 7.5 with 3mM Sodium Azide | |
56654 | |
Mouse | |
IgG1 | |
Primary | |
100μg | |
Human,Mouse,Rat |
Western Blot | |
Unconjugated | |
Purified | |
CAB-, CAB-1, CAB1CAB, DKFZp586J0523, neural proliferation differentiation and control protein 1, neural proliferation, differentiation and control, 1, NPDC-1 | |
NPDC1 | |
Hybridoma produced by the fusion of splenocytes from mice immunized with a synthetic peptide derived from human NPDC-1 protein and mouse myeloma cells. | |
Monoclonal | |
Ammonium sulfate precipitation | |
Store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title