NPEC-caged-serotonin, Tocris™

Caged serotonin

Manufacturer: R&D Systems 3991/50

Quantity 50mg

 View more versions of this product

Catalog No. 39-915-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C19H19N3O5
    Formula Weight 369.37
    Synonym 3-((N)-1-(2-Nitrophenyl)ethylcarboxy)-(2-Aminoethyl)-1H-indol-5-ol
    Chemical Name or Material NPEC-caged-serotonin

    Specifications

    Formula Weight 369.37
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C19H19N3O5
    CAS 1257326-22-9
    Synonym 3-((N)-1-(2-Nitrophenyl)ethylcarboxy)-(2-Aminoethyl)-1H-indol-5-ol
    Recommended Storage Store at -20°C
    Chemical Name or Material NPEC-caged-serotonin

    Store at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit