Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPEPL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530821
Description
Recommended positive controls: IMR32, MCF-7. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (87%), Xenopus laevis (93%), Pig (95%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probably catalyzes the removal of unsubstituted N-terminal amino acids from various peptides.
Specifications
NPEPL1 | |
Polyclonal | |
Unconjugated | |
NPEPL1 | |
Aminopeptidase-like 1; bA261P9.2; BC023239; C85514; KIAA1974; mKIAA1974; NPEPL1; Probable aminopeptidase NPEPL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79716 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NDH3 | |
NPEPL1 | |
Recombinant fragment corresponding to a region within amino acids 220 and 463 of Human NPEPL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction