Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPEPPS Recombinant Rabbit Monoclonal Antibody (HL1531)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547084
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene encodes the puromycin-sensitive aminopeptidase, a zinc metallopeptidase which hydrolyzes amino acids from the N-terminus of its substrate. The protein has been localized to both the cytoplasm and to cellular membranes. This enzyme degrades enkaphalins in the brain, and studies in mouse suggest that it is involved in proteolytic events regulating the cell cycle.
Specifications
NPEPPS | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P55786, Q11011 | |
NPEPPS | |
Recombinant protein encompassing a sequence within the center region of human NPEPPS. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Feline, Zebrafish | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
HL1531 | |
Unconjugated | |
NPEPPS | |
AAP-S; aminopeptidase puromycin sensitive; cytosol alanyl aminopeptidase; goku; metalloproteinase MP100; MP100; NPEPPS; Psa; Puromycin-sensitive aminopeptidase; R74825 | |
Rabbit | |
Protein A | |
RUO | |
101098097, 19155, 407726, 480538, 50558, 9520 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction