Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPHP1 Mouse anti-Human, Unconjugated, Clone: 1M4, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen NPHP1
Applications Western Blot, Immunofluorescence
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS21851MI
View Documents
MilliporeSigma
MABS2185100UL
100 μL
Each for $372.00
Add to cart
 
MABS21852MI
View Documents
MilliporeSigma
MABS218525UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

NPHP1 Monoclonal antibody specifically detects NPHP1 in Human samples. It is validated for Western Blotting, Immunofluorescence.
Specifications

Specifications

NPHP1
Unconjugated
Mouse
Nephrocystin-1;Juvenile nephronophthisis 1 protein
A synthetic polypeptide fused by 16 different NPHP1 peptide sequences predicted to have the highest antigenicity. The sequence was converted to cDNA and the polypeptide was expressed and purified from E. coli.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot, Immunofluorescence
Purified
Human
NPHP1, NPH1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit