Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPHP3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548439

Catalog No. PA548439

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with zebrafish and xenopus based on sequence homology.

Required for normal ciliary development and function. Inhibits disheveled-1-induced canonical Wnt-signaling activity and may also play a role in the control of non-canonical Wnt signaling which regulates planar cell polarity. Probably acts as a molecular switch between different Wnt signaling pathways. Required for proper convergent extension cell movements.
Specifications

Specifications

NPHP3
Polyclonal
PBS with 0.09% sodium azide
Q7Z494
Nephrocystin-3, NPHP3, KIAA2000
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27031
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NPHP3
Liquid
NPHP3
KLH conjugated synthetic peptide between 144-173 amino acids from the N-terminal region of human NPHP3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit