Learn More
NPHP5/IQCB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 157471AP
Catalog No. 50-173-0449
Add to cart
Description
Description
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
NPHP5/IQCB1
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15051, Q8BP00
IQ motif containing B1, IQCB1, KIAA0036, Nephrocystin 5, NPHP5, PIQ, SLSN5
Rabbit
IgG
150 μL
-20°C
Primary
303915, 320299, 9657
Immunocytochemistry, Immunofluorescence
Unconjugated
IQCB1
Liquid
Iqcb1
NPHP5,IQCB1 Fusion Protein Ag8203
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat