Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551219
Description
The antibody detects endogenous levels of total NPL protein.
Catalyzes the cleavage of N-acetylneuraminic acid (sialic acid) to form pyruvate and N-acetylmannosamine via a Schiff base intermediate. It prevents sialic acids from being recycled and returning to the cell surface. Involved in the N- glycolylneuraminic acid (Neu5Gc) degradation pathway. Although human is not able to catalyze formation of Neu5Gc due to the inactive CMAHP enzyme, Neu5Gc is present in food and must be degraded.
Specifications
NPL | |
Polyclonal | |
Unconjugated | |
NPL | |
0610033B02Rik; C112; C1orf13; dihydrodipicolinate synthetase homolog 1; N-acetylneuraminate lyase; N-acetylneuraminate pyruvate lyase; N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase); N-acetylneuraminate pyruvate-lyase; N-acetylneuraminic acid aldolase; NAL; NALase; Npl; NPL1; Sialate lyase; sialate-pyruvate lyase; Sialic acid aldolase; sialic acid lyase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80896 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BXD5 | |
NPL | |
Fusion protein of human NPL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction