Learn More
Aviva Systems Biology Corp NPM2 Antibody - N-terminal region (ARP58311_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58311_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NPM2. Alternate names: MGC78655ApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-62281