Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557603
Description
Immunogen sequence: ARNHDHQEIA VPVANLKLSC QPMLSLDDFQ LQPPVTFRLK SGSGPVRITG RHQIVTMSND VSE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
The protein encoded by this gene is related to the nuclear chaperone phosphoproteins, nucleoplasmin and nucleophosmin. This protein is strongly expressed in diverse cell types where it localizes primarily to the nucleus. Based on its similarity to nucleoplasmin and nucleophosmin, this protein likely functions as a molecular chaperone in the cell nucleus.
Specifications
NPM3 | |
Polyclonal | |
Unconjugated | |
NPM3 | |
Npm3; Nub1; nucleophosmin/nucleoplasmin 3; nucleophosmin/nucleoplasmin family, member 3; nucleophosmin/nucleoplasmin, 3; nucleoplasmin 3; nucleoplasmin-3; PORMIN; RGD1565542; TMEM123 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75607 | |
NPM3 | |
Recombinant protein corresponding to Human NPM3. Recombinant protein control fragment (Product #RP-97050). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction