Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPR3 Mouse anti-Human, Clone: OTI2B12, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA501045

Catalog No. 50-167-2188


Add to cart

Description

Description

ANPC is a receptor for atrial natriuretic peptide. It does not exhibit guanylate cyclase activity. There seem to be at least three ANP receptors: two with guanylate cyclase activity (ANPA and ANPB) and one (ANPC) which is probably responsible for the clearance of ANP from the circulation without a role in signal transduction.
Specifications

Specifications

NPR3
Monoclonal
0.72 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P17342
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4883
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2B12
Unconjugated
NPR3
Liquid
NPR3
Full length human recombit protein of human NPR3 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit