Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPRL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120959

This item is not returnable.
View return policy
Description
Positive test controls include: HL-60, U-937, Mouse brain.
NPRL2 Polyclonal antibody specifically detects NPRL2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
NPRL2 | |
Polyclonal | |
Unconjugated | |
NPRL2 | |
2810446G01Rik; G21; G21 protein; GATOR complex protein NPRL2; Gene 21 protein; homologous to yeast nitrogen permease (candidate tumor suppressor); nitrogen permase homolog; nitrogen permease regulator 2-like protein; nitrogen permease regulator-like 2; nitrogen permease regulator-like 2 (S. cerevisiae); NPR 2L; NPR L2; NPR like 2; NPR2; NPR2 like; NPR2L; NPR2-like protein; NPR2-like, GATOR1 complex subunit; NPRL 2; Nprl2; Tumor suppressor candidate 4; TUSC 4; TUSC 4 protein; TUSC4; TUSC4 protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human NPRL2 (NP_006536.3). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
3.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WTW4, Q9WUE4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10641, 363138, 56032 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title