NPS™ Spilfyter™ Cellulose Sorbent Spill Kits

Quick spill control for large, non-aggressive spills

Manufacturer: NPS™ 152020

 View more versions of this product

Catalog No. 19-066-496

  Check Availability
Add to Cart

    Specifications

    Capacity (English) Sorbing 19 gal.
    Capacity (Metric) Sorbing 72L
    Capacity 20 gal., 75.7L
    Includes Four 3 in. x 4 ft. cellulose socks, 24 8 x 18 in. cellulose pads, disposal bag and twist tie, 20 gal. poly drum with Lever-lock metal ring top
    Material Cellulose
    Capacity (Metric) 75.7L
    Product Type NPS Spill Kit
    Capacity (English) 20 gal.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit