Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPTN Mouse anti-Canine, Human, Mouse, Clone: OTI2D4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504330

Catalog No. 50-167-5048


Add to cart

Description

Description

Neuroplastin encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

NPTN
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P97300, Q9Y639
GP55, GP65, SDFR1, SDR1, np55, np65
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
20320, 27020, 487637
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2D4
Unconjugated
NPTN
Liquid
NPTN
Full length human recombit protein of human NPTN produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit