Tocris™ NPY 5RA972

Potent and selective NPY Y5 antagonist

Manufacturer: Tocris™ 3677/10

 View more versions of this product

Catalog No. 36-771-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors NPY 5RA972
    Purity >98
    Molecular Weight 351.44
    Quantity 10mg
    Product Type NPY 5RA972

    Potent and selective neuropeptide Y Y5 receptor antagonist (IC50 values are 3.1, >10000, >10000 and >10000nM for Y5, Y1, Y2 and Y4 receptors respectively). Does not block NPY-induced feeding in vivo. Orally active and brain penetrant.

    • Chemical Name: N-[4-Methyl-9-(1-methylethyl)-9H-carbazol-3-yl]-4-morpholinecarboxamide
    • Formula: C21H25N3O2
    • CAS Number: 439861-56-0
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit