Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPY Y2 Receptor-C/N Terminus Rabbit anti-Mouse, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$271.56 - $491.04

Specifications

Antigen NPY Y2 Receptor-C/N Terminus
Concentration 1 mg/mL
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0048 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA14136100UL
100 μL
Each for $491.04
Add to cart
 
50-210-0049 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA1413650UL
50 μL
Each for $271.56
Add to cart
 
Description

Description

NPY Y2 Receptor-C/N Terminus Polyclonal antibody specifically detects NPY Y2 Receptor-C/N Terminus in Mouse, Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

NPY Y2 Receptor-C/N Terminus
Immunohistochemistry
Unconjugated
Rabbit
Mouse, Rat
P97295
18167, 66024
IgG
Primary
-20° C, Avoid Freeze/Thaw Cycles
NPY2R
1 mg/mL
Polyclonal
Liquid, Purified
RUO
PBS with 0.05% sodium azide
MGC124241; MGC124242; MGC124244; Neuropeptide Y receptor type 2; neuropeptide Y receptor Y2; neuropeptide Y receptor Y2-like; neuropeptide Y/peptide YY-Y2 receptor; Neuropeptide y-y2; neuropeptide Y-Y2 receptor; Npy2r; NPY2-R; NPY-Y2 receptor; Y2 receptor
(C) GAEDENQTVEVKVE-NH2 and (C) TDFSEATNV-COOH
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit