Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPY Y2 Receptor Rabbit anti-Mouse, Rat, Polyclonal, Neuromics™
Rabbit Polyclonal Antibody
Supplier: Neuromics, Inc RA1411250UL

Description
NPY Y2 Receptor Polyclonal antibody specifically detects NPY Y2 Receptor in Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, ImmunohistochemistrySpecifications
NPY Y2 Receptor | |
Polyclonal | |
Unconjugated | |
NPY2R | |
MGC124241; MGC124242; MGC124244; Neuropeptide Y receptor type 2; neuropeptide Y receptor Y2; neuropeptide Y receptor Y2-like; neuropeptide Y/peptide YY-Y2 receptor; Neuropeptide y-y2; neuropeptide Y-Y2 receptor; Npy2r; NPY2-R; NPY-Y2 receptor; Y2 receptor | |
(C)TDSFSEATNV. Cys added for conjugation to KLH | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
1.14 mg/mL | |
PBS with 50% glycerol and 0.05% sodium azide | |
P97295 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18167, 66024 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction