Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPY2R Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | NPY2R |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126679
|
Novus Biologicals
NBP310889100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
NPY2R Polyclonal specifically detects NPY2R in Human samples. It is validated for Western Blot.Specifications
NPY2R | |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Neuroscience, Neurotransmission | |
PBS buffer, 2% sucrose | |
4887 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
neuropeptide Y receptor type 2, neuropeptide Y receptor Y2, NPY2-R, NPY-Y2 receptor, Y2 receptor | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human NPY2R (NP_000901). Peptide sequence PIGAEADENQTVEEMKVEQYGPQTTPRGELVPDPEPELIDSTKLIEVQVV | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title