Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NQO1 Monoclonal Antibody (A180), DyLight™ 650
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA516098
Description
NQO1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), Flow
NQO1 is a member of the NAD(P)H dehydrogenase (quinone) family and a cytoplasmic 2-electron reductase. This FAD-binding protein forms homodimers and reduces quinones to hydroquinones. NQO1's enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species. Altered expression of the protein has been seen in many tumors and is also associated with Alzheimer's disease (AD).
Specifications
NQO1 | |
Monoclonal | |
0.72 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
P05982, P15559, Q64669 | |
NQO1 | |
Full length recombinant NQO1 (from human lung). | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
A180 | |
DyLight 650 | |
NQO1 | |
AV001255; Azoreductase; DHQU; DIA4; Diaphorase (NADH/NADPH); diaphorase (NADH/NADPH) (cytochrome b-5 reductase); Diaphorase 4; diaphorase 4 (NADH/NADPH); diaphorase-4; dioxin-inducible 1; DTD; DT-diaphorase; menadione reductase; NAD(P)H dehydrogenase (quinone); NAD(P)H dehydrogenase [quinone] 1; NAD(P)H dehydrogenase, quinone 1; NAD(P)H menadione oxidoreductase 1, dioxin inducible; NAD(P)H quinone dehydrogenase 1; NAD(P)H: quinone oxidoreductase 1; NAD(P)H:menadione oxidoreductase; NAD(P)H:menadione oxidoreductase 1; NAD(P)H:Quinone acceptor oxidoreductase type 1; NAD(P)H:quinone oxidoreductase 1; NAD(P)H:quinone oxireductase; NADPH-d; nicotinamide adenine dinucleotide phosphate diaphorase; Nmo1; Nmo-1; Nmor1; NMORI; NQ01; NQO1; Ox1; Ox-1; phylloquinone reductase; QR1; quinone reductase 1 | |
Mouse | |
Protein G | |
RUO | |
1728, 18104, 24314, 610935 | |
4°C, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction