Learn More
Aviva Systems Biology Corp NR0B1 Antibody - middle region (ARP48005_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR0B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NR0B1. Alternate names: AHC, AHCH, AHX, DAX-1, DAX1, DSS, GTD, HHG, NROB1, SRXY2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59701