Learn More
Aviva Systems Biology Corp NR0B1 Antibody - N-terminal region (ARP48004_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48004_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NR0B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR0B1. Alternate names: AHC, AHCH, AHX, DAX-1, DAX1, DSS, GTD, HHG, NROB1, SRXY2ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-59700