Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR0B2 Recombinant Protein, Abnova

Manufacturer:  Abnova Corporation H00008431P0125

 View more versions of this product

Catalog No. 89-011-258

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH30207
Solution
8431
NR0B2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NR0B2
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
54.01
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSTSQPGACPCQGAASRPAILYALLSSSLKAVPRPRSRCLCRQHRPVQLCAPHRTCREALDVLAKTVAFLRNLPSFWQLPPQDQRRLLQGCWGPLFLLGLAQDAVTFEVAEAPVPSILKKILLEEPSSSGGSGQLPDRPQPSLAAVQWLQCCLESFWSLELSPKEYACLKGTILFNPDVPGLQAASHIGHLQQEAHWVLCEVLEPWCPAAQGRLTRVLLTASTLKSIPTSLLGDLFFRPIIGDVDIAGLLGDMLLLR
FLJ17090|SHP|SHP1
NR0B2
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit