Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR1H4 Mouse anti-Human, Clone: OTI4F12, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF809306
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The steroid receptor superfamily acts through direct association with DNA sequences known as hormone response elements (HREs) and bind DNA as either homo- or heterodimers. The promiscuous mediator of heterodimerization, RXR, is the receptor for 9-cis retinoic acid, and dimerizes with VDR, TR, PPAR, as well as several novel receptors including LXR (also referred to as RLD-1) and FXR. FXR and LXR fall into a category of proteins termed orphan receptors because of their lack of a defined function, and in the case of LXR, the lack of a defined ligand. FXR has been shown to bind a class of lipid molecules called farnesoids. LXR/RXR heterodimers have highest affinity for DR-4 DNA elements while FXR/RXR heterodimers bind IR-1 elements. Both LXR/RXR and FXR/RXR heterodimers retain their responsiveness to 9-cis retinoic acid.Specifications
NR1H4 | |
Monoclonal | |
Unconjugated | |
NR1H4 | |
Lyophilized | |
NR1H4 | |
Human recombit protein fragment corresponding to amino acids 21-297 of human NR1H4 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4F12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96RI1 | |
BAR, FXR, HRR-1, HRR1, PFIC5, RIP14 | |
Mouse | |
IgG1 | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
9971 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title