Learn More
NR1I3 Mouse anti-Human, Clone: OTI8E3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805367
Catalog No. 50-166-7400
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the nuclear receptor superfamily, and is a key regulator of xenobiotic and endobiotic metabolism. The protein binds to DNA as a monomer or a heterodimer with the retinoid X receptor and regulates the transcription of target genes involved in drug metabolism and bilirubin clearance, such as cytochrome P450 family members. Unlike most nuclear receptors, this transcriptional regulator is constitutively active in the absence of ligand but is regulated by both agonists and inverse agonists. Ligand binding results in translocation of this protein to the nucleus, where it activates or represses target gene transcription. These ligands include bilirubin, a variety of foreign compounds, steroid hormones, and prescription drugs. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
NR1I3
Monoclonal
Unconjugated
NR1I3
Lyophilized
NR1I3
Human recombit protein fragment corresponding to amino acids 1-236 of human NR1I3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8E3
PBS with 8% trehalose and no preservative; pH 7.3
Q14994
CAR, CAR1, MB67
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9970