Learn More
NR1I3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 210421AP
Catalog No. 50-173-3223
Add to cart
Description
Description
This gene encodes a member of the nuclear receptor superfamily, and is a key regulator of xenobiotic and endobiotic metabolism. The protein binds to DNA as a monomer or a heterodimer with the retinoid X receptor and regulates the transcription of target genes involved in drug metabolism and bilirubin clearance, such as cytochrome P450 family members. Unlike most nuclear receptors, this transcriptional regulator is constitutively active in the absence of ligand but is regulated by both agonists and inverse agonists.
Specifications
Specifications
NR1I3
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35627, Q14994, Q9QUS1
CAR, CAR BETA, CAR1, Constitutive active response, MB67, NR1I3, Orphan nuclear receptor MB67
Rabbit
IgG
150 μL
-20°C
Primary
12355, 65035, 9970
Western Blot
Unconjugated
NR1I3
Liquid
Nr1i3
NR1I3 Fusion Protein Ag15378
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat