Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR2C2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 209811AP150UL
Description
The antibody is specific to NR2C2.
Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.Specifications
NR2C2 | |
Polyclonal | |
Unconjugated | |
NR2C2 | |
hTAK1, NR2C2, Orphan nuclear receptor TAK1, Orphan nuclear receptor TR4, TAK1, Testicular receptor 4, TR2R1, TR4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22026, 7182 | |
-20°C | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49116, P49117 | |
NR2C2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction