Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2C2 Mouse anti-Human, Clone: OTI2H2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526854

Catalog No. PIMA526854


Add to cart

Description

Description

Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes (Yoshikawa et al., 1996 [PubMed 8661150]).
Specifications

Specifications

NR2C2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P49116
TAK1, TR2R1, TR4, hTAK1, Nuclear hormone receptor TR4, TR4 nuclear hormone receptor, nuclear receptor subfamily 2 group C member 2, orphan nuclear receptor TAK1, orphan nuclear receptor TR4, testicular nuclear receptor 4, testicular receptor 4
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7182
Immunohistochemistry (Paraffin), Western Blot
OTI2H2
Unconjugated
NR2C2
Liquid
NR2C2
Human recombit protein fragment corresponding to amino acids 1-241 of NR2C2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit