Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR2C2AP Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548416
Description
Predicted to react with bovine and mouse based on sequence homology.
May act as a repressor of NR2C2-mediated transactivation by suppressing the binding between NR2C2/TR4 and the TR4-response element in target genes.
Specifications
NR2C2AP | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q86WQ0 | |
NR2C2AP | |
KLH conjugated synthetic peptide between 8-37 amino acids from the N-terminal region of human NR2C2AP. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NR2C2AP | |
Nuclear receptor 2C2-associated protein, TR4 orphan receptor-associated 16 kDa protein, NR2C2AP, TRA16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126382 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction