Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR3A Mouse anti-Mouse,Rat, Unconjugated, Clone: N416/40, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen NR3A
Applications Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN18651MI
View Documents
MilliporeSigma
MABN1865100UG
100 μg
Each for $372.00
Add to cart
 
MABN18652MI
View Documents
MilliporeSigma
MABN186525UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

NR3A Monoclonal antibody specifically detects NR3A in Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

NR3A
Unconjugated
Mouse
NR3A;NMDAR3A;N-methyl-D-aspartate receptor subtype 3A
Linear peptide from the N-terminal region of mouse Glutamate receptor ionotropic, NMDA3A (NR3A).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin)
Purified
Mouse, Rat
Grin3a
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit