Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR5A1 Rabbit anti-Human, Human, Mouse, Rat, Clone: ARC2457, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Manufacturer:  Thermo Scientific MA537904

Encompass

Catalog No. PIMA537904


Add to cart

Description

Description

Positive Samples: BxPC-3, Mouse ovary, Mouse spleen

NR5A1 Recombinant Monoclonal antibody specifically detects NR5A1 in Human, Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

NR5A1
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P33242, P50569, Q13285
AD4BP; Ad4BP/SF-1; adrenal 4 binding protein; adrenal 4-binding protein; ELP; ELP-3; Embryonal long terminal repeat-binding protein; embryonal LTR-binding protein; FTZ1; FTZF1; Ftz-F1; fushi tarazu 1 factor homolog; fushi tarazu factor homolog 1; hSF-1; Nr5a1; nuclear receptor AdBP4; nuclear receptor subfamily 5 group A member 1; nuclear receptor subfamily 5, group A, member 1; POF7; SF1; SF-1; SPGF8; SRXY3; steroid hormone receptor Ad4BP; Steroid hydroxylase positive regulator; steroidogenic factor 1; steroidogenic factor 1 nuclear receptor; steroidogenic factor-1; STF-1
Recombinant protein of human STF1
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Western Blot
ARC2457
Unconjugated
Nr5a1
Liquid
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2516, 26423, 83826
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit