Learn More
Aviva Systems Biology Corp NR5A2 ANTIBODY 100UG/100UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP38386_T100
This is a rabbit polyclonal antibody against NR5A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NR5A2. Alternate names: B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, hB1F, hB1F-2, LRH1ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Pig, Rat, Sheep
Catalog No. 50-014-53104