Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR6/CLC Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0186-11D28, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212421AF350

Catalog No. NP212421A30


Add to cart

Description

Description

NR6/CLC Monoclonal antibody specifically detects NR6/CLC in Human samples. It is validated for Western Blot.
Specifications

Specifications

NR6/CLC
MM0186-11D28
Western Blot
B-cell stimulating factor 3, B-cell stimulatory factor 3, BSF3BSF-3, cardiotrophin-like cytokine factor 1, CISS2, CLCNNT-1, CRLF1 associated cytokine-like factor 1, neurotrophin-1/B-cell stimulating factor-3, NNT1B-cell-stimulating factor 3, Novel neurotrophin-1, NR6
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CLCF1
Human
0.1 mL
Primary
23529
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit