Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLC Mouse anti-Human, Alexa Fluor 750, Clone: MM0186-11D28, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212421AF750

Catalog No. NP212421A70


Add to cart

Description

Description

CLC Monoclonal antibody specifically detects CLC in Human samples. It is validated for Western Blot.
Specifications

Specifications

CLC
MM0186-11D28
Western Blot
Conjugated, Purified
CLCF1
Human
Protein G purified
Apoptosis
Primary
23529.0
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
B-cell stimulating factor 3, B-cell stimulatory factor 3, BSF3BSF-3, cardiotrophin-like cytokine factor 1, CISS2, CLCNNT-1, CRLF1 associated cytokine-like factor 1, neurotrophin-1/B-cell stimulating factor-3, NNT1B-cell-stimulating factor 3, Novel neurotrophin-1, NR6
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit