Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NRAMP1/SLC11A1 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8400S100UG
This item is not returnable.
View return policy
Description
NRAMP1/SLC11A1 Monoclonal specifically detects NRAMP1/SLC11A1 in Human samples. It is validated for Flow Cytometry.Specifications
NRAMP1/SLC11A1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_000569 | |
SLC11A1 | |
NS0 mouse myeloma cell line transfected with human NRAMP1/SLC11A1 Met1-Gly550 Accession # NP_000569 | |
RUO | |
6556 | |
Human | |
IgG2b |
Flow Cytometry | |
910825 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
natural resistance-associated macrophage protein 1, LSH, member 1, NRAMP 1, NRAMP1solute carrier family 11 (sodium/phosphate symporters), member 1, NRAMPnatural resistance-associated macrophage protein 1, SLC11A1, solute carrier family 11 (proton-coupled divalent metal ion transporters) | |
Mouse | |
100 μg | |
Primary | |
Detects HEK293 human embryonic kidney cell line transfected with human NRAMP1/SLC11A1 by Flow Cytometry. Does not detect untransfected or irrelevant transfected HEK293 cells. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction