Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NRARP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555248
Description
Immunogen sequence: MSQAELSTCS APQTQRIFQE AVRKGNTQEL QSLLQNMTNC EFNVNSFGPE GQTALHQSVI DGNLELVKLL VKFGADIRLA NRDGWSALHI AAF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Specifications
NRARP | |
Polyclonal | |
Unconjugated | |
Nrarp | |
2700054M22Rik; Maternal Effect Lethal family member (mel-11)-like; NOTCH-regulated ankyrin repeat protein; notch-regulated ankyrin repeat-containing protein; NRARP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441478 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6K4 | |
Nrarp | |
Recombinant protein corresponding to Human NRARP. Recombinant protein control fragment (Product #RP-93100). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction