Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRBP Mouse anti-Human, Non-human primate, Clone: OTI5C3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524925

Catalog No. PIMA524925


Add to cart

Description

Description

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications

Specifications

NRBP
Monoclonal
1.48 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q4R8X0, Q9UHY1
BCON3, MADM, MUDPNP, NRBP, multiple domain putative nuclear protein, myeloid leukemia factor 1 adaptor molecule, nuclear receptor-binding protein
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
101866221, 29959
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
OTI5C3
Unconjugated
NRBP1
Liquid
NRBP1
Full-length protein expressed in 293T cell transfected with human NRBP1 expression vector
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit