Learn More
NRBP Mouse anti-Canine, Human, Non-human primate, Rat, Clone: OTI7E2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524928
Catalog No. PIMA524928
Add to cart
Description
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
Specifications
NRBP
Monoclonal
1.1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UHY1, Q4R8X0
BCON3, MADM, MUDPNP, NRBP, multiple domain putative nuclear protein, myeloid leukemia factor 1 adaptor molecule, nuclear receptor-binding protein
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
101866221, 29959, 475704, 619579
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
OTI7E2
Unconjugated
NRBP1
Liquid
NRBP1
Full-length protein expressed in 293T cell transfected with human NRBP1 expression vector
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Non-human primate, Rat