Learn More
NrCAM Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S364-51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242191V
Catalog No. NBP242191V
Add to cart
Description
Description
NrCAM Monoclonal antibody specifically detects NrCAM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
NrCAM
S364-51
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
NRCAM
Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM. Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ~50% identity with Neurofascin.
Protein G purified
Store at 4C in the dark.
Monoclonal
Approx 50% identity with NeurofascinDetects approx 160kDa.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM
Mouse
IgG2a
0.1 ml
Primary
4897
Human, Mouse, Rat