Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NrCAM Mouse anti-Human, Mouse, Rat, HRP, Clone: S364-51, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242191H

Catalog No. NBP242191H


Add to cart

Description

Description

NrCAM Monoclonal antibody specifically detects NrCAM in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

NrCAM
S364-51
Western Blot
Purified
NRCAM
Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM. Rat: 96% identity (795/822 amino acids identical). Human: 91% identity (753/822 amino acids identical) ~50% identity with Neurofascin.
Protein G purified
Store at 4C in the dark.
Monoclonal
Approx 50% identity with NeurofascinDetects approx 160kDa.
Western Blot
HRP
PBS with No Preservative
bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM
Mouse
IgG2a
0.1 ml
Primary
4897
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit