Learn More
Nrf1 Mouse anti-Human, Clone: C.15200013, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259255
Catalog No. NBP259255
Add to cart
Description
Description
Nrf1 Monoclonal antibody specifically detects Nrf1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
Specifications
Nrf1
C.15200013
Western Blot 1:1000, Chromatin Immunoprecipitation 2 ug
Purified
NRF1
The exact immunogen is propietary information.
Protein A purified
RUO
Primary
4899
Western Blot, ChIP assay
Unconjugated
PBS with 0.05% Sodium Azide
Alpha palindromic-binding protein, ALPHA-PAL, EWG, NRF-1, nuclear respiratory factor 1
Mouse
IgG2a
50 ug
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human