Learn More
Medchemexpress LLC Nrf2 activator-4 | 2383016-68-8 | 98.03% | 468.90 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14608610MM/1ML
Encompass_Preferred
Nrf2 activator-4 (Compound 20a) is a highly potent, orally active Nrf2 activator with an EC50 of 0.63 μM. It suppresses reactive oxygen species against oxidative stress in microglia and effectively recovers learning and memory impairment in a scopolamine-induced mouse model.
  • Highly potent and orally active Nrf2 activator
  • EC50 of 0.63 μM
  • Suppresses reactive oxygen species against oxidative stress in microglia
  • Effectively recovers learning and memory impairment in a scopolamine-induced mouse model
  • Light yellow to yellow solid appearance
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-003-60712