Learn More
Medchemexpress LLC Nrf2-IN-1 | 1610022-76-8 | 99.9% | 383.87 | C21H22ClN3O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10102510MM 1ML
Encompass_Preferred
Nrf2-IN-1 is a small-molecule inhibitor of nuclear factor-erythroid 2-related factor 2 (Nrf2) that targets the Keap1-Nrf2 pathway and is developed for preclinical research in acute myeloid leukemia (AML). It is available as a powder and as a ready-to-use 10 mM solution in DMSO, with reported activity inhibiting Nrf2 activation and inducing apoptosis in AML models.
  • Inhibits Nrf2 activation in vitro.
  • Reduces proliferation and induces apoptosis in AML cell lines.
  • Supplied as powder and as a 10 mM solution in DMSO (1 mL), ready for use.
  • High reported purity and characterized molecular properties.
  • Soluble in DMSO; recommended storage conditions provided.
Catalog No. 50-002-12039