Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NRG1 beta-1 Human ELISA Kit Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Sandwich ELISA Kit

$494.00

Specifications

Storage Requirements 2°C to 8°C
Assay Sensitivity 50 pg/mL
Assay Range 82.3-20,000 pg/mL
Test Time 1 hr. 20 min.
Sample Volume Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHNRG1 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
EHNRG1
96 Tests
Pack for $494.00
Add to cart
 
Description

Description

The Human NRG1beta-1 (Hu NRG1) ELISA quantitates Hu NRG1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu NRG1. Principle of the method The Human NRG1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Neuregulin-1 beta 1/NRG1 beta 1 is one of many soluble and transmembrane NRG1 isoforms. All family members share an EGF-like domain that interacts with the ErbB family of tyrosine kinase receptors. NRG1 isoforms can be classified into type I (neu differentiation factor, NDF; heregulin, HRG; acetylcholine receptor inducing activity, ARIA), type II (glial growth factor, GGF) and type III (sensory and motor neuron-derived factor, SMDF) isoforms. NRG1 regulates multiple nervous system functions including neurotransmitter expression, axon guidance, synapse formation and plasticity, glial cell development, and axon myelination. It regulates heart morphogenesis and contractility and also plays a cardioprotective role following tissue injury. Multiple polymorphisms and aberrant expression of NRG1 isoforms are associated with the development of schizophrenia and many cancers.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

2°C to 8°C
82.3-20,000 pg/mL
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
<10%
50 pg/mL
1 hr. 20 min.
<12%
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit