Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NRG4 Monoclonal Antibody (C2)
Mouse Monoclonal Antibody
$243.00 - $588.50
Specifications
Antigen | NRG4 |
---|---|
Clone | C2 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541647
|
Invitrogen™
MA541647 |
200 μL |
Each for $588.50
|
|
|||||
PIMA543644
|
Invitrogen™
MA543644 |
20 μL |
Each for $243.00
|
|
|||||
Description
NRG4 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
The neuregulins, including NRG4, activate type-1 growth factor receptors to initiating cell-to-cell signaling through tyrosine phosphorylation.Specifications
NRG4 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q8WWG1 | |
145957 | |
Recombinant protein Neuregulin 4. The antigen corresponds to amino acid range 1-62 of the target protein. | |
Antibody |
C2 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
NRG4 | |
AI552600; heregulin 4; HRG4; neuregulin 4; Neuregulin-4; NRG4; NRG-4; Pro-neuregulin-4, membrane-bound isoform; pro-NRG4 | |
NRG4 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title