Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRG4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550341

Catalog No. PA550341


Add to cart

Description

Description

The antibody detects endogenous levels of total NRG4 protein.

The neuregulins, including NRG4, activate type-1 growth factor receptors to initiating cell-to-cell signaling through tyrosine phosphorylation.
Specifications

Specifications

NRG4
Polyclonal
Unconjugated
NRG4
Liquid
NRG4
Fusion protein corresponding to residues near the N terminal of human neuregulin 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.4 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q8WWG1
HRG4
Rabbit
IgG
100 μL
-20°C
Primary
145957
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit