Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NRIP Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130075C

Catalog No. NBP130075C


Add to cart

Description

Description

NRIP Polyclonal antibody specifically detects NRIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

NRIP
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
1200006M05Rik, Androgen receptor complex-associated protein, ARCAP, DDB1 and CUL4 associated factor 6, DDB1- and CUL4-associated factor 6, FLJ23798, IQ motif and WD repeat-containing protein 1, IQ motif and WD repeats 1, IQWD1, MSTP055, NRIP, Nuclear receptor interaction protein, PC326
Rabbit
IgG
100 ul
Store at 4C in the dark.
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified
DCAF6
Maps to a region between residue 300 and 350 of human nuclear receptor interaction protein (IQ motif and WD repeats 1) using the numbering given in entry NP_001017977.1
Immunogen affinity purified
RUO
Primary
55827
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only