Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NRIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559209
Description
Immunogen sequence: KLGSSKDMQP HNILQRRLME TNLSKLRSGP RVPWASKTNK LNQAKSEGLK KSEEDDMILV SCQCAGKDVK A Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.
NRIP3 is a protein coding gene.
Specifications
| NRIP3 | |
| Polyclonal | |
| Unconjugated | |
| Nrip3 | |
| A330103B05Rik; C11orf14; D7H11orf14; ICRFP703B1614Q5.2; ICRFP703N2430Q5.2; NRIP3; nuclear receptor interacting protein 3; nuclear receptor-interacting protein 3; NY-SAR-105; Sarcoma antigen NY-SAR-105 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56675 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NQ35 | |
| Nrip3 | |
| Recombinant protein corresponding to Human NRIP3. Recombinant protein control fragment (Product #RP-97500). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction