Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRIP3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309282100UL
Description
NRIP3 Polyclonal specifically detects NRIP3 in Human samples. It is validated for Western Blot.Specifications
NRIP3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
C11orf14, chromosome 11 open reading frame 14, nuclear receptor interacting protein 3, nuclear receptor-interacting protein 3, NY-SAR-105, Sarcoma antigen NY-SAR-105 | |
The immunogen is a synthetic peptide directed towards the middle region of human NRIP3 (NP_065696). Peptide sequence EKNLSLGLQTLRSLKCIINLDKHRLIMGKTDKEEIPFVETVSLNEDNTSE | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
56675 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction