Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NRL Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2822930.1ML
Description
NRL Polyclonal specifically detects NRL in Human samples. It is validated for Western Blot.Specifications
NRL | |
Polyclonal | |
PBS, 2% sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4901 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
D14S46Eneural retina-specific leucine zipper protein, neural retina leucine zipper, NRL-MAF, RP27neural retinal-specific leucine zipper | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human NRL | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction